Assay Details
Target Gene Details
Entrez Gene ID: | 9025 |
Gene Name: | ring finger protein 8 |
Gene Aliases: |
hRNF8 |
Location: |
Chr.6:37353972-37394738 on Build GRCh38 |
Assay Gene Location: | Within Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF8 | NM_003958.3 | 3 | 736 | NP_003949.1 |
| NM_183078.2 | 3 | 736 | NP_898901.1 | |
| NR_046399.1 | 3 | 842 | ||
| XM_006715241.2 | 3 | 736 | XP_006715304.1 | |
| XM_006715242.2 | 3 | 736 | XP_006715305.1 | |
| XM_017011462.1 | 3 | 587 | XP_016866951.1 | |
| XM_017011463.1 | 3 | 592 | XP_016866952.1 | |
| XM_017011464.1 | 3 | 741 | XP_016866953.1 | |
| AB014546.1 | 3 | 655 | ||
| AF334675.1 | 3 | 543 | AAQ14887.1 | |
| AK022075.1 | 3 | 637 | ||
| AK222765.1 | 3 | 654 | BAD96485.1 | |
| AK298319.1 | 3 | 760 | ||
| AK308840.1 | 3 | 643 | ||
| BC007517.2 | 3 | 665 | AAH07517.1 | |
| BT007446.1 | 3 | 543 | AAP36114.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1015689 | Chr.6:36550916 - 39052434 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map