Assay Details
Target Gene Details
Entrez Gene ID: | 284340 |
Gene Name: | C-X-C motif chemokine ligand 17 |
Gene Aliases: |
DMC, Dcip1, UNQ473, VCC-1, VCC1 |
Location: |
Chr.19:42428538-42443048 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CXCL17 | NM_198477.2 | NP_940879.1 | ||
| NR_133910.1 | ||||
| AK292985.1 | ||||
| AY358433.1 | AAQ88799.1 | |||
| AY598464.1 | AAU04875.1 | |||
| BC093946.1 | AAH93946.1 | |||
| BC112095.1 | AAI12096.1 | |||
| BM978124.1 | ||||
| BU684368.1 |
Target Gene Details
Entrez Gene ID: | 100996307 |
Gene Name: | LIPE antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.19:42397148-42652355 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LIPE-AS1 | NR_073179.1 | |||
| NR_073180.1 | ||||
| AK096849.1 | BAC04875.1 | |||
| AK310497.1 | ||||
| AK311181.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526186 | Chr.19:42172329 - 42699355 on Build GRCh38 | Gain |
|
| esv2422471 | Chr.19:42228989 - 43165333 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map