Assay Details
Target Gene Details
Entrez Gene ID: | 79140 |
Gene Name: | coiled-coil domain containing 28B |
Gene Aliases: |
- |
Location: |
Chr.1:32200386-32205390 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCDC28B | NM_001301011.1 | NP_001287940.1 | ||
| NM_024296.4 | NP_077272.2 | |||
| XM_011542113.2 | XP_011540415.1 | |||
| XM_017002307.1 | XP_016857796.1 | |||
| XM_017002308.1 | XP_016857797.1 | |||
| XM_017002309.1 | XP_016857798.1 | |||
| AK225113.1 | ||||
| AK291904.1 | ||||
| BC022848.2 | AAH22848.1 | |||
| BM981930.1 |
Target Gene Details
Entrez Gene ID: | 56063 |
Gene Name: | transmembrane protein 234 |
Gene Aliases: |
AASL548, C1orf91, PRO1105, RP4-622L5, dJ622L5.7 |
Location: |
Chr.1:32204807-32222371 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM234 | XM_011541795.2 | 10 | 3271 | XP_011540097.1 |
| XM_017001814.1 | 10 | 2018 | XP_016857303.1 | |
| XM_017001815.1 | 10 | 2350 | XP_016857304.1 | |
| XM_017001817.1 | 9 | 3196 | XP_016857306.1 | |
| XM_017001818.1 | 10 | 3094 | XP_016857307.1 | |
| XM_017001819.1 | 12 | 2572 | XP_016857308.1 | |
| XM_017001821.1 | 12 | 2240 | XP_016857310.1 | |
| XM_017001827.1 | 10 | 3341 | XP_016857316.1 | |
| XM_017001828.1 | 11 | 2611 | XP_016857317.1 |
More Information
Set Membership: |
|

Genomic Map