Assay Details
Target Gene Details
Entrez Gene ID: | 1824 |
Gene Name: | desmocollin 2 |
Gene Aliases: |
ARVD11, CDHF2, DG2, DGII/III, DSC3 |
Location: |
Chr.18:31065974-31102432 on Build GRCh38 |
Assay Gene Location: | Within Exon 18 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DSC2 | NM_004949.4 | 17 | 3809 | NP_004940.1 |
| NM_024422.4 | 16 | 3763 | NP_077740.1 | |
| XM_005258206.4 | 16 | 3264 | XP_005258263.1 | |
| BC063291.1 | 16 | 3745 | AAH63291.1 | |
| BE813598.1 | ||||
| BX648425.1 | CAH56152.1 | |||
| CR749226.1 | 16 | 3482 | CAH18082.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2244 | Chr.18:31055588 - 31086414 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map