Assay Details
Target Gene Details
Entrez Gene ID: | 10330 |
Gene Name: | canopy FGF signaling regulator 2 |
Gene Aliases: |
HP10390, MSAP, TMEM4, ZSIG9 |
Location: |
Chr.12:56310428-56316344 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CNPY2 | NM_014255.5 | NP_055070.1 | ||
| AB015631.1 | BAA76498.1 | |||
| AK312921.1 | ||||
| AY032624.1 | AAK38148.1 | |||
| AY359102.1 | AAQ89460.1 | |||
| BC065015.1 | AAH65015.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv482992 | Chr.12:56206217 - 57706217 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map