Assay Details
Target Gene Details
Entrez Gene ID: | 84678 |
Gene Name: | lysine demethylase 2B |
Gene Aliases: |
CXXC2, FBXL10, Fbl10, JHDM1B, PCCX2 |
Location: |
Chr.12:121402289-121582268 on Build GRCh38 |
Assay Gene Location: | Within Intron 26 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KDM2B | XM_011538867.2 | XP_011537169.1 |
Target Gene Details
Entrez Gene ID: | 80196 |
Gene Name: | ring finger protein 34 |
Gene Aliases: |
CARP-1, CARP1, RFI, RIF, RIFF, hRFI |
Location: |
Chr.12:121400083-121424353 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 6 - Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RNF34 | NM_001256858.1 | NP_001243787.1 | ||
| NM_025126.3 | NP_079402.2 | |||
| NM_194271.2 | NP_919247.1 | |||
| AB084914.1 | BAC11802.1 | |||
| AF306709.1 | AAK51328.1 | |||
| AK025439.1 | BAB15132.1 | |||
| AK096994.1 | ||||
| AK130355.1 | ||||
| AK225285.1 | ||||
| AY098934.1 | AAM29180.1 | |||
| BC007826.2 | AAH07826.1 | |||
| BI559962.1 | ||||
| BT007283.1 | AAP35947.1 | |||
| CR457342.1 | CAG33623.1 |
More Information
Set Membership: |
|

Genomic Map