Assay Details
Target Gene Details
Entrez Gene ID: | 60401 |
Gene Name: | ectodysplasin A2 receptor |
Gene Aliases: |
EDA-A2R, EDAA2R, TNFRSF27, XEDAR |
Location: |
Chr.X:66594384-66639303 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 8 - Exon 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2305e212 | Chr.X:66550620 - 66717352 on Build GRCh38 | Gain |
|
| nsv520630 | Chr.X:66486237 - 66797185 on Build GRCh38 | Gain+Loss |
|
| esv3576919 | Chr.X:65837678 - 66804698 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map