Assay Details
Target Gene Details
Entrez Gene ID: | 90411 |
Gene Name: | multiple coagulation factor deficiency 2 |
Gene Aliases: |
F5F8D, F5F8D2, LMAN1IP, SDNSF |
Location: |
Chr.2:46901870-46941855 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MCFD2 | NM_001171506.2 | 5 | 3552 | NP_001164977.1 |
| NM_001171507.2 | 4 | 3444 | NP_001164978.1 | |
| NM_001171508.2 | 4 | 3596 | NP_001164979.1 | |
| NM_001171509.2 | 3 | 3303 | NP_001164980.1 | |
| NM_001171510.2 | 3 | 3289 | NP_001164981.1 | |
| NM_001171511.2 | 3 | 3441 | NP_001164982.1 | |
| NM_139279.5 | 4 | 3458 | NP_644808.1 | |
| AF537214.1 | 4 | 3395 | AAP23162.1 | |
| AK095006.1 | 1 | 2165 | ||
| AL833900.1 | 4 | 3393 | CAD38756.1 | |
| BC037845.1 | 3 | 3201 | AAH37845.1 | |
| BC040357.2 | 4 | 3384 | AAH40357.1 | |
| CR749562.1 | 3 | 3246 | CAH18359.1 | |
| M23161.1 | 2 | 3127 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv457352 | Chr.2:46777649 - 47098834 on Build GRCh38 | Loss |
|
| nsv1004828 | Chr.2:46829710 - 47587343 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map