Assay Details
Target Gene Details
Entrez Gene ID: | 253512 |
Gene Name: | solute carrier family 25 member 30 |
Gene Aliases: |
KMCP1 |
Location: |
Chr.13:45393316-45434016 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 11 - Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC25A30 | NM_001010875.3 | NP_001010875.1 | ||
| NM_001286806.1 | NP_001273735.1 | |||
| NM_001286807.1 | NP_001273736.1 | |||
| XM_005266321.2 | XP_005266378.1 | |||
| XM_006719793.3 | XP_006719856.1 | |||
| XM_006719794.1 | XP_006719857.1 | |||
| XM_011535026.1 | XP_011533328.1 | |||
| XM_011535027.2 | XP_011533329.1 | |||
| XM_017020523.1 | XP_016876012.1 | |||
| XM_017020524.1 | XP_016876013.1 | |||
| AK094133.1 | ||||
| AK095465.1 | ||||
| AK302967.1 | ||||
| AL832206.1 | 4 | 2026 | ||
| BC132739.1 | ||||
| BC136760.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv223e201 | Chr.13:45377870 - 46464067 on Build GRCh38 | Deletion |
|
| nsv510579 | Chr.13:45305226 - 45424938 on Build GRCh38 | Deletion |
|
| esv3631956 | Chr.13:45391109 - 45405451 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map