Assay Details
Target Gene Details
Entrez Gene ID: | 148252 |
Gene Name: | DIRAS family GTPase 1 |
Gene Aliases: |
Di-Ras1, GBTS1, RIG |
Location: |
Chr.19:2714567-2721392 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DIRAS1 | NM_145173.3 | 2 | 698 | NP_660156.1 |
| XM_011527713.2 | 2 | 863 | XP_011526015.1 | |
| AB076888.1 | 1 | 639 | BAC01115.1 | |
| AK124445.1 | ||||
| AY056037.1 | 1 | 540 | AAL23715.1 | |
| AY059641.1 | 1 | 540 | AAL17968.1 | |
| AY180973.1 | 1 | 540 | AAO22153.1 | |
| BC030660.1 | 2 | 677 | AAH30660.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv828399 | Chr.19:2714966 - 2724570 on Build GRCh38 | Gain |
|
| nsv578398 | Chr.19:2701508 - 2717882 on Build GRCh38 | Gain |
|
| nsv509714 | Chr.19:2519629 - 2794403 on Build GRCh38 | Insertion |
|
| nsv470109 | Chr.19:2557612 - 2719369 on Build GRCh38 | Loss |
|
| nsv828397 | Chr.19:2688987 - 2800506 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map