Assay Details
Target Gene Details
Entrez Gene ID: | 572 |
Gene Name: | BCL2 associated agonist of cell death |
Gene Aliases: |
BBC2, BCL2L8 |
Location: |
Chr.11:64269828-64284704 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| BAD | NM_004322.3 | 1 | 444 | NP_004313.1 |
| NM_032989.2 | 2 | 190 | NP_116784.1 | |
| AB451254.1 | 1 | 108 | ||
| AB451378.1 | 1 | 108 | ||
| AF021792.1 | 1 | 108 | AAB72092.1 | |
| AF031523.1 | 1 | 108 | AAB88124.1 | |
| AK291863.1 | 2 | 190 | ||
| AK303051.1 | 2 | 190 | ||
| AK309150.1 | 2 | 190 | ||
| BC001901.2 | 1 | 116 | AAH01901.1 | |
| BC095431.1 | 2 | 155 | AAH95431.1 | |
| BE255791.1 | 1 | 349 | ||
| BG435829.1 | 1 | 380 | ||
| BG748336.1 | 2 | 178 | ||
| BT006678.1 | 1 | 108 | AAP35324.1 | |
| CR541935.1 | 1 | 108 | CAG46733.1 | |
| CR541959.1 | 1 | 108 | CAG46757.1 | |
| U66879.1 | 1 | 174 | AAB36516.1 |
Target Gene Details
Entrez Gene ID: | 56834 |
Gene Name: | G protein-coupled receptor 137 |
Gene Aliases: |
C11orf4, GPR137A, TM7SF1L1 |
Location: |
Chr.11:64270337-64289500 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPR137 | XM_011545168.2 | XP_011543470.1 | ||
| XM_011545170.2 | XP_011543472.1 | |||
| XM_011545171.2 | XP_011543473.1 | |||
| XM_011545172.2 | XP_011543474.1 | |||
| XM_017018016.1 | XP_016873505.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss |
|
| nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
|
| nsv469961 | Chr.11:64055468 - 64318495 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map