Assay Details
Target Gene Details
Entrez Gene ID: | 27297 |
Gene Name: | CGRP receptor component |
Gene Aliases: |
CGRP-RCP, CGRPRCP, RCP, RCP9 |
Location: |
Chr.7:66114818-66154568 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CRCP | NM_001040647.1 | 5 | 1862 | NP_001035737.1 |
| NM_001040648.1 | 3 | 1730 | NP_001035738.1 | |
| NM_001142414.1 | 5 | 1806 | NP_001135886.1 | |
| NM_014478.4 | 6 | 1961 | NP_055293.1 | |
| NR_024548.1 | 7 | 2083 | ||
| AK023295.1 | 1 | 1056 | ||
| BX647867.1 | CAI46069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2761079 | Chr.7:65601260 - 66224746 on Build GRCh38 | Gain+Loss |
|
| esv2674673 | Chr.7:66151430 - 66155249 on Build GRCh38 | Deletion |
|
| esv2664145 | Chr.7:65814760 - 66395282 on Build GRCh38 | Deletion |
|
| esv3613502 | Chr.7:66151382 - 66155221 on Build GRCh38 | Loss |
|
| nsv1021186 | Chr.7:66027614 - 66198451 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map