Assay Details
Target Gene Details
Entrez Gene ID: | 639 |
Gene Name: | PR domain 1 |
Gene Aliases: |
BLIMP1, PRDI-BF1 |
Location: |
Chr.6:106046729-106109939 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDM1 | NM_001198.3 | 1 | 203 | NP_001189.2 |
| XM_017011187.1 | XP_016866676.1 | |||
| AF084199.1 | AAC33300.1 | |||
| AK289556.1 | 1 | 147 | ||
| AL832963.1 | 1 | 137 | CAH56276.1 | |
| AY198414.1 | 1 | 203 | AAO45623.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1025231 | Chr.6:106003137 - 106186514 on Build GRCh38 | Gain |
|
| dgv6134n100 | Chr.6:106081902 - 106464172 on Build GRCh38 | Gain |
|
| nsv830755 | Chr.6:106013830 - 106176327 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map