Assay Details
Target Gene Details
Entrez Gene ID: | 55805 |
Gene Name: | LRP2 binding protein |
Gene Aliases: |
- |
Location: |
Chr.4:185363878-185397160 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LRP2BP | NM_018409.3 | 8 | 2105 | NP_060879.2 |
| XM_005263125.4 | 7 | 1385 | XP_005263182.1 | |
| XM_006714260.3 | 9 | 2435 | XP_006714323.1 | |
| XM_006714265.3 | 8 | 1495 | XP_006714328.1 | |
| XM_011532110.2 | 8 | 2308 | XP_011530412.1 | |
| XM_011532111.2 | 8 | 1358 | XP_011530413.1 | |
| XM_017008410.1 | 9 | 3318 | XP_016863899.1 | |
| XM_017008411.1 | 8 | 1923 | XP_016863900.1 | |
| AB037746.1 | 8 | 2105 | ||
| BC150161.1 | 8 | 1612 | ||
| BC152444.1 | 8 | 2105 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428456 | Chr.4:185328666 - 185492417 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map