Assay Details
Target Gene Details
Entrez Gene ID: | 81558 |
Gene Name: | family with sequence similarity 117 member A |
Gene Aliases: |
- |
Location: |
Chr.17:49710325-49764156 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM117A | NM_030802.3 | 8 | 1563 | NP_110429.1 |
| XM_017025182.1 | 8 | 2321 | XP_016880671.1 | |
| AF123073.1 | 8 | 1524 | AAK01477.1 | |
| AK302376.1 | 8 | 1377 | ||
| BC037572.1 | 8 | 1575 | AAH37572.1 | |
| BC065199.1 | 8 | 1528 | AAH65199.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv525469 | Chr.17:49704315 - 49720153 on Build GRCh38 | Gain |
|

Genomic Map