Assay Details
Target Gene Details
Entrez Gene ID: | 10456 |
Gene Name: | HCLS1 associated protein X-1 |
Gene Aliases: |
HCLSBP1, HS1BP1, SCN3 |
Location: |
Chr.1:154272563-154275875 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HAX1 | NM_001018837.1 | NP_001018238.1 | ||
| NM_006118.3 | NP_006109.2 | |||
| AK290626.1 | ||||
| AK300676.1 | ||||
| AK310000.1 | ||||
| AK311450.1 | ||||
| BC005240.1 | AAH05240.1 | |||
| BC014314.1 | AAH14314.1 | |||
| BC015209.1 | AAH15209.1 | |||
| BC016730.1 | AAH16730.1 | |||
| EU190982.1 | ||||
| EU190983.1 | ||||
| U68566.1 | AAB51196.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map