Assay Details
Target Gene Details
Entrez Gene ID: | 5991 |
Gene Name: | regulatory factor X3 |
Gene Aliases: |
- |
Location: |
Chr.9:3218297-3526001 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 24 - Intron 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RFX3 | NM_001282116.1 | NP_001269045.1 | ||
| NM_134428.2 | NP_602304.1 | |||
| XM_006716840.1 | XP_006716903.1 | |||
| XM_006716843.1 | XP_006716906.1 | |||
| XM_006716844.1 | XP_006716907.1 | |||
| XM_006716845.2 | XP_006716908.1 | |||
| XM_006716847.3 | XP_006716910.1 | |||
| XM_011517998.1 | XP_011516300.1 | |||
| XM_011517999.1 | XP_011516301.1 | |||
| XM_011518000.1 | XP_011516302.1 | |||
| XM_011518001.1 | XP_011516303.1 | |||
| XM_017015000.1 | XP_016870489.1 | |||
| XM_017015001.1 | XP_016870490.1 | |||
| XM_017015002.1 | XP_016870491.1 | |||
| AK289540.1 | ||||
| BC022191.2 | AAH22191.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1017474 | Chr.9:3019068 - 3247215 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map