Assay Details
Target Gene Details
Entrez Gene ID: | 2961 |
Gene Name: | general transcription factor IIE subunit 2 |
Gene Aliases: |
FE, TF2E2, TFIIE-B, TTD6 |
Location: |
Chr.8:30578514-30658241 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GTF2E2 | NM_002095.4 | 7 | 967 | NP_002086.1 |
| XM_017013363.1 | 7 | 827 | XP_016868852.1 | |
| XM_017013364.1 | 7 | 757 | XP_016868853.1 | |
| BC030572.2 | 7 | 961 | AAH30572.1 | |
| S67861.1 | 7 | 904 | AAB20414.1 | |
| X63469.1 | 7 | 956 | CAA45069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526509 | Chr.8:30328056 - 30643198 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map