Assay Details
Target Gene Details
Entrez Gene ID: | 57560 |
Gene Name: | intraflagellar transport 80 |
Gene Aliases: |
ATD2, SRTD2, WDR56 |
Location: |
Chr.3:160256986-160399532 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IFT80 | NM_001190241.1 | 1 | 130 | NP_001177170.1 |
| NM_020800.2 | 1 | 130 | NP_065851.1 | |
| AK125342.1 | 1 | 130 | ||
| AK303410.1 | 1 | 128 |
Target Gene Details
Entrez Gene ID: | 10051 |
Gene Name: | structural maintenance of chromosomes 4 |
Gene Aliases: |
CAP-C, CAPC, SMC-4, SMC4L1 |
Location: |
Chr.3:160399304-160434962 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SMC4 | NM_001002800.2 | 1 | 100 | NP_001002800.1 |
| NM_001288753.1 | 1 | 100 | NP_001275682.1 | |
| DB444342.2 | 1 | 100 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv822305 | Chr.3:160399357 - 160399807 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map