Assay Details
Target Gene Details
Entrez Gene ID: | 84456 |
Gene Name: | l(3)mbt-like 3 (Drosophila) |
Gene Aliases: |
MBT-1, MBT1 |
Location: |
Chr.6:130018583-130141449 on Build GRCh38 |
Assay Gene Location: | Within Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| L3MBTL3 | NM_001007102.2 | 22 | 2403 | NP_001007103.1 |
| NM_032438.2 | 23 | 2478 | NP_115814.1 | |
| XM_005267161.4 | 23 | 2654 | XP_005267218.1 | |
| XM_006715576.3 | 23 | 2583 | XP_006715639.1 | |
| XM_006715578.3 | 23 | 2462 | XP_006715641.1 | |
| XM_011536179.2 | 23 | 2461 | XP_011534481.1 | |
| XM_011536180.2 | 23 | 2480 | XP_011534482.1 | |
| XM_011536181.2 | 23 | 2415 | XP_011534483.1 | |
| XM_011536183.2 | 23 | 3011 | XP_011534485.1 | |
| XM_011536184.2 | 23 | 2415 | XP_011534486.1 | |
| XM_017011355.1 | 22 | 2494 | XP_016866844.1 | |
| XM_017011356.1 | 22 | 2386 | XP_016866845.1 | |
| AB058701.1 | 22 | 2376 | ||
| AK092756.1 | 13 | 1655 | ||
| AK094137.1 | 23 | 2485 | ||
| AK290219.1 | 22 | 2395 | ||
| BC060845.1 | 22 | 2397 | AAH60845.1 |
Target Gene Details
Entrez Gene ID: | 154075 |
Gene Name: | sterile alpha motif domain containing 3 |
Gene Aliases: |
- |
Location: |
Chr.6:130133203-130365425 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SAMD3 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv604663 | Chr.6:129482340 - 130671010 on Build GRCh38 | Gain |
|
| esv3610781 | Chr.6:130108614 - 130242086 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map