Assay Details
Target Gene Details
Entrez Gene ID: | 23581 |
Gene Name: | caspase 14 |
Gene Aliases: |
- |
Location: |
Chr.19:15049480-15058293 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CASP14 | NM_012114.2 | 3 | 324 | NP_036246.1 |
| XM_011527861.1 | 3 | 324 | XP_011526163.1 | |
| AF097874.1 | 2 | 160 | AAD16173.1 | |
| AK313992.1 | 3 | 324 | ||
| BC069541.1 | 2 | 160 | AAH69541.1 | |
| BC103868.1 | 2 | 160 | AAI03869.1 | |
| BC103869.1 | 2 | 160 | AAI03870.1 | |
| EU262664.1 | 2 | 160 | ||
| EU502748.1 | 3 | 420 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv654e214 | Chr.19:15050284 - 15101244 on Build GRCh38 | Gain |
|
| esv3643762 | Chr.19:15052933 - 15067870 on Build GRCh38 | Gain |
|
| esv2758746 | Chr.19:14986138 - 15148368 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map