Assay Details
Target Gene Details
Entrez Gene ID: | 94005 |
Gene Name: | phosphatidylinositol glycan anchor biosynthesis class S |
Gene Aliases: |
- |
Location: |
Chr.17:28553388-28571869 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PIGS | NM_033198.3 | 6 | 973 | NP_149975.1 |
| AB057723.1 | 6 | 600 | BAB60853.1 | |
| AK055512.1 | BAB70938.1 | |||
| AK075428.1 | 6 | 628 | BAC11615.1 | |
| AK297046.1 | 5 | 492 | ||
| AK314583.1 | 6 | 636 | ||
| AY359112.1 | 5 | 868 | AAQ89470.1 | |
| BC007301.1 | 4 | 357 | ||
| BC069228.1 | 6 | 623 | AAH69228.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1064087 | Chr.17:28462253 - 28755692 on Build GRCh38 | Gain |
|
| nsv1064958 | Chr.17:28490519 - 28565886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map