Assay Details
Target Gene Details
Entrez Gene ID: | 5367 |
Gene Name: | pro-melanin concentrating hormone |
Gene Aliases: |
MCH, ppMCH |
Location: |
Chr.12:102196459-102199540 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PMCH | NM_002674.3 | 1 | 54 | NP_002665.2 |
| XM_017019483.1 | 1 | 1749 | XP_016874972.1 | |
| BC018048.1 | AAH18048.1 | |||
| CB047053.1 | 1 | 63 | ||
| M57703.1 | AAA63214.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv560033 | Chr.12:102163421 - 102233833 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map