Assay Details
Target Gene Details
Entrez Gene ID: | 7818 |
Gene Name: | death associated protein 3 |
Gene Aliases: |
DAP-3, MRP-S29, MRPS29, bMRP-10 |
Location: |
Chr.1:155688104-155739010 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 9 - Intron 9 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33869 | Chr.1:155253492 - 155948170 on Build GRCh38 | Loss |
|
| nsv428246 | Chr.1:155588406 - 155816040 on Build GRCh38 | Gain |
|
| esv2718429 | Chr.1:155648347 - 155763436 on Build GRCh38 | Deletion |
|
| nsv3065 | Chr.1:155712977 - 155757733 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map