Assay Details
Target Gene Details
Entrez Gene ID: | 5165 |
Gene Name: | pyruvate dehydrogenase kinase 3 |
Gene Aliases: |
CMTX6, GS1-358P8.4 |
Location: |
Chr.X:24465227-24550466 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 2 - Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PDK3 | NM_001142386.2 | NP_001135858.1 | ||
| NM_005391.4 | NP_005382.1 | |||
| AK301965.1 | ||||
| BC015948.1 | AAH15948.1 | |||
| BG717206.1 | ||||
| BI549903.1 | ||||
| L42452.1 | AAC42011.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv2240e212 | Chr.X:24487194 - 24496632 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map