Assay Details
Target Gene Details
Entrez Gene ID: | 81025 |
Gene Name: | gap junction protein alpha 9 |
Gene Aliases: |
CX58, CX59, GJA10 |
Location: |
Chr.1:38874067-38881626 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA9 | NM_030772.4 | 2 | 1884 | NP_110399.2 |
| AF271261.1 | 1 | 1847 | AAK55516.1 | |
| BU689816.1 | 1 | 457 |
Target Gene Details
Entrez Gene ID: | 100527950 |
Gene Name: | GJA9-MYCBP readthrough |
Gene Aliases: |
- |
Location: |
Chr.1:38862490-38881626 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJA9-MYCBP | NR_037633.1 | 2 | 1884 | |
| NR_037634.1 | ||||
| NR_037635.1 | ||||
| NR_037636.1 | ||||
| NR_037637.1 | ||||
| AK123051.1 | ||||
| FY210901.1 | ||||
| FY211253.1 | ||||
| FY211295.1 |
Target Gene Details
Entrez Gene ID: | 105378663 |
Gene Name: | uncharacterized LOC105378663 |
Gene Aliases: |
- |
Location: |
Chr.1:38873401-38876226 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC105378663 | NR_135048.1 | |||
| BX115242.1 | ||||
| HY234287.1 |
More Information
Set Membership: |
|

Genomic Map