Assay Details
Target Gene Details
Entrez Gene ID: | 55832 |
Gene Name: | cullin associated and neddylation dissociated 1 |
Gene Aliases: |
TIP120, TIP120A |
Location: |
Chr.12:67269281-67314692 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CAND1 | NM_018448.4 | 1 | 168 | NP_060918.2 |
| AB020636.2 | 1 | 70 | ||
| AK096010.1 | 1 | 74 | ||
| AK292456.1 | 1 | 114 | ||
| AK314358.1 | 1 | 56 | ||
| AL136810.1 | 1 | 80 | CAB66744.1 | |
| DB064887.1 | 1 | 168 | ||
| DB098973.1 | 1 | 145 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3629822 | Chr.12:67262967 - 67357107 on Build GRCh38 | Loss |
|
| esv3629823 | Chr.12:67263258 - 67312978 on Build GRCh38 | Loss |
|
| nsv1054111 | Chr.12:67163000 - 67471762 on Build GRCh38 | Gain |
|
| nsv559212 | Chr.12:66979769 - 67457145 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map