Assay Details
Target Gene Details
Entrez Gene ID: | 126669 |
Gene Name: | Src homology 2 domain containing E |
Gene Aliases: |
- |
Location: |
Chr.1:154469637-154502059 on Build GRCh38 |
Assay Gene Location: | Within Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SHE | NM_001010846.2 | 6 | 2661 | NP_001010846.1 |
| NR_135169.1 | 6 | 2656 | ||
| XM_005244891.4 | XP_005244948.1 | |||
| XM_011509163.2 | XP_011507465.1 | |||
| AK074067.1 | 7 | 2080 | BAB84893.1 | |
| AK126006.1 | 4 | 1926 | ||
| CR936667.1 | 6 | 2104 | ||
| CR936736.1 | 3 | 3336 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv521887 | Chr.1:154264094 - 154711630 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map