Assay Details
Target Gene Details
Entrez Gene ID: | 124857 |
Gene Name: | WAP, follistatin/kazal, immunoglobulin, kunitz and netrin domain containing 2 |
Gene Aliases: |
GASP-1, WFDC20B, WFIKKNRP, hGASP-1 |
Location: |
Chr.17:50834650-50842353 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WFIKKN2 | NM_175575.5 | 2 | 2178 | NP_783165.1 |
| XM_017024172.1 | 2 | 1657 | XP_016879661.1 | |
| AF468657.1 | 2 | 1500 | AAL77058.1 | |
| AK127743.1 | 2 | 1838 | ||
| AK298172.1 | 2 | 1657 | ||
| AK312821.1 | 2 | 1672 | ||
| AY358142.1 | 2 | 2013 | AAQ88509.1 | |
| BC140842.1 | 2 | 1572 | ||
| BC140844.1 | 2 | 1572 |
More Information
Set Membership: |
|

Genomic Map