Assay Details
Target Gene Details
| Entrez Gene ID: | 4610 | 
| Gene Name: | v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog | 
| Gene Aliases: | L-Myc, LMYC, MYCL1, bHLHe38 | 
| Location: | Chr.1:39895424-39902015 on Build GRCh38 | 
| Assay Gene Location: | Within Exon 1 | 
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID | 
|---|---|---|---|---|
| MYCL | NM_001033081.2 | NP_001028253.1 | ||
| NM_001033082.2 | NP_001028254.2 | |||
| NM_005376.4 | 2 | 1222 | NP_005367.2 | |
| AK296078.1 | ||||
| AK296838.1 | ||||
| BC011864.2 | 2 | 1110 | AAH11864.1 | |
| HQ628638.1 | 
Target Copy Number Variation Details
| DGV Version: | Release date: 2016-05-15, GRCh GRCh38 | 
| Target Variation | Location | CNV Subtype | Genes | 
|---|---|---|---|
| esv2758933 | Chr.1:39759471 - 40194760 on Build GRCh38 | Loss |  LOC101929536  MYCL  TRIT1  BMP8B  PPT1  MFSD2A  OXCT2  PPIE  CAP1  RLF | 
 Back To Top
 Back To TopMore Information
| Set Membership: |  Intragenic  Exonic  DGV Variation | 
Panther Classification:
Gene Ontology Categories:
 Back To Top
 Back To Top
 Genomic Map
Genomic Map Back To Top
 Back To Top