Assay Details
Target Gene Details
Entrez Gene ID: | 83450 |
Gene Name: | dynein regulatory complex subunit 3 |
Gene Aliases: |
CFAP134, LRRC48 |
Location: |
Chr.17:17972766-18016889 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DRC3 | NM_001130090.1 | 10 | 1337 | NP_001123562.1 |
| NM_001130091.1 | 10 | 1337 | NP_001123563.1 | |
| NM_001130092.1 | 9 | 1248 | NP_001123564.1 | |
| NM_031294.3 | 9 | 1248 | NP_112584.3 | |
| XM_006721583.3 | 7 | 1032 | XP_006721646.1 | |
| XM_011524018.2 | 10 | 1397 | XP_011522320.1 | |
| XM_011524019.2 | 10 | 1243 | XP_011522321.1 | |
| XM_011524020.2 | 10 | 1522 | XP_011522322.1 | |
| XM_011524021.2 | 10 | 1338 | XP_011522323.1 | |
| XM_011524022.2 | 10 | 1404 | XP_011522324.1 | |
| XM_011524023.2 | 10 | 1404 | XP_011522325.1 | |
| XM_011524025.2 | 4 | 1664 | XP_011522327.1 | |
| XM_011524026.2 | 4 | 439 | XP_011522328.1 | |
| AK093317.1 | 8 | 1099 | ||
| AK293011.1 | 10 | 1216 | ||
| AL136926.1 | 8 | 2546 | CAB66860.2 | |
| BC040276.1 | 9 | 1124 | AAH40276.1 | |
| BC047873.1 | 9 | 1176 | AAH47873.1 | |
| BC050419.1 | 10 | 1198 | AAH50419.1 | |
| BC050665.1 | 10 | 1194 | AAH50665.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510701 | Chr.17:17947160 - 18052171 on Build GRCh38 | Deletion |
|
| nsv1132448 | Chr.17:17996686 - 18000386 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map