Assay Details
Target Gene Details
Entrez Gene ID: | 4314 |
Gene Name: | matrix metallopeptidase 3 |
Gene Aliases: |
CHDS6, MMP-3, SL-1, STMY, STMY1, STR1 |
Location: |
Chr.11:102835797-102843689 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MMP3 | NM_002422.4 | 10 | 1640 | NP_002413.1 |
| AI628953.1 | 1 | 267 | ||
| AK223283.1 | 10 | 1558 | BAD97003.1 | |
| AK223291.1 | 10 | 1559 | BAD97011.1 | |
| BC069676.1 | 10 | 1560 | AAH69676.1 | |
| BC069716.1 | 10 | 1560 | AAH69716.1 | |
| BC074815.2 | AAH74815.1 | |||
| BC074869.2 | AAH74869.1 | |||
| BC105954.1 | 10 | 1553 | AAI05955.1 | |
| BC107490.1 | 10 | 1553 | AAI07491.1 | |
| BC107491.1 | 10 | 1553 | AAI07492.1 | |
| X05232.1 | 10 | 1540 | CAA28859.1 |
Target Gene Details
Entrez Gene ID: | 100288077 |
Gene Name: | Wilms tumor 1 associated protein pseudogene 1 |
Gene Aliases: |
- |
Location: |
Chr.11:102783676-102836766 on Build GRCh38 |
Assay Gene Location: | Within Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| WTAPP1 | NR_038390.1 | |||
| AK097805.1 |
More Information
Set Membership: |
|

Genomic Map