Assay Details
Target Gene Details
Entrez Gene ID: | 130872 |
Gene Name: | AHA1, activator of heat shock 90kDa protein ATPase homolog 2 (yeast) |
Gene Aliases: |
Hch1 |
Location: |
Chr.2:61177418-61191210 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| AHSA2 | XM_011532541.2 | 8 | 3564 | XP_011530843.1 |
| XM_011532543.2 | 9 | 3848 | XP_011530845.1 | |
| XM_011532545.2 | 8 | 2272 | XP_011530847.1 | |
| XM_017003377.1 | 7 | 2170 | XP_016858866.1 | |
| XM_017003378.1 | 9 | 3523 | XP_016858867.1 |
Target Gene Details
Entrez Gene ID: | 9736 |
Gene Name: | ubiquitin specific peptidase 34 |
Gene Aliases: |
- |
Location: |
Chr.2:61187455-61470714 on Build GRCh38 |
Assay Gene Location: | Within Exon 79 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| USP34 | NM_014709.3 | 79 | 9943 | NP_055524.3 |
| AB011142.2 | 80 | 9998 | BAA25496.2 | |
| AB018272.1 | 22 | 2872 | BAA34449.1 | |
| AJ586138.1 | 77 | 9468 | CAE51938.1 | |
| AK001193.1 | 19 | 2534 | ||
| AK022637.1 | 9 | 983 | ||
| AK023171.1 | 11 | 1220 | ||
| AK125898.1 | 38 | 4462 | ||
| AL050092.1 | 14 | 2039 | CAB43264.1 | |
| AL050376.1 | 1 | 1009 | ||
| BC022783.1 | 16 | 2242 | AAH22783.1 | |
| BC062325.1 | 7 | 697 | AAH62325.1 | |
| BC107761.1 | 6 | 608 | AAI07762.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2760616 | Chr.2:61136348 - 61200572 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map