Assay Details
Target Gene Details
Entrez Gene ID: | 149647 |
Gene Name: | family with sequence similarity 71 member A |
Gene Aliases: |
- |
Location: |
Chr.1:212624447-212626778 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM71A | NM_153606.3 | 1 | 958 | NP_705834.2 |
| AK057358.1 | 1 | 571 | ||
| AK301981.1 | ||||
| AK315021.1 | 1 | 582 | ||
| BC035007.1 | 1 | 933 | AAH35007.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv549139 | Chr.1:212158952 - 212703560 on Build GRCh38 | Gain |
|
| esv2758996 | Chr.1:212474801 - 212669761 on Build GRCh38 | Loss |
|

Genomic Map