Assay Details
Target Gene Details
Entrez Gene ID: | 9538 |
Gene Name: | EI24, autophagy associated transmembrane protein |
Gene Aliases: |
EPG4, PIG8, TP53I8 |
Location: |
Chr.11:125569216-125584689 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| EI24 | NM_001290135.1 | NP_001277064.1 | ||
| NM_004879.4 | NP_004870.3 | |||
| NR_110769.1 | ||||
| NR_110770.1 | ||||
| XM_011543069.1 | XP_011541371.1 | |||
| XM_011543070.1 | XP_011541372.1 | |||
| XM_017018582.1 | XP_016874071.1 | |||
| AF010313.2 | AAC39531.2 | |||
| BQ936285.1 | ||||
| DB520736.1 | ||||
| DB567734.1 |
Target Gene Details
Entrez Gene ID: | 105369550 |
Gene Name: | STT3A antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.11:125568108-125592568 on Build GRCh38 |
Assay Gene Location: | Within Intron 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STT3A-AS1 | NR_132372.1 | |||
| DB453285.1 | ||||
| DB470444.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv983073 | Chr.11:125583612 - 125585567 on Build GRCh38 | Duplication |
|
| esv32964 | Chr.11:125495069 - 126046387 on Build GRCh38 | Gain+Loss |
|
More Information
Set Membership: |
|

Genomic Map