Assay Details
Target Gene Details
Entrez Gene ID: | 84279 |
Gene Name: | protease associated domain containing 1 |
Gene Aliases: |
C2orf7, PAP21 |
Location: |
Chr.2:73228006-73233228 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRADC1 | NM_032319.1 | NP_115695.1 | ||
| AK312077.1 | ||||
| AY358605.1 | AAQ88968.1 | |||
| BC005069.1 | AAH05069.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv2776 | Chr.2:73204726 - 73255176 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map