Assay Details
Target Gene Details
Entrez Gene ID: | 5320 |
Gene Name: | phospholipase A2 group IIA |
Gene Aliases: |
MOM1, PLA2, PLA2B, PLA2L, PLA2S, PLAS1, sPLA2 |
Location: |
Chr.1:19975431-19980439 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PLA2G2A | NM_000300.3 | NP_000291.1 | ||
| NM_001161727.1 | NP_001155199.1 | |||
| NM_001161728.1 | NP_001155200.1 | |||
| BC005919.1 | AAH05919.1 | |||
| BX641714.1 | ||||
| CB132375.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv527818 | Chr.1:19979394 - 20053652 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map