Assay Details
Target Gene Details
Entrez Gene ID: | 2974 |
Gene Name: | guanylate cyclase 1 soluble subunit beta 2 (pseudogene) |
Gene Aliases: |
GC-SB2 |
Location: |
Chr.13:50994511-51066157 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 7 - Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GUCY1B2 | NR_003923.2 | |||
| AF038499.2 | AAD09440.2 | |||
| AK296746.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv561618 | Chr.13:49907577 - 51521449 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map