Assay Details
Target Gene Details
Entrez Gene ID: | 25801 |
Gene Name: | grancalcin |
Gene Aliases: |
GCL |
Location: |
Chr.2:162318768-162371605 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GCA | XM_005246446.2 | 8 | 929 | XP_005246503.1 |
| XM_006712398.3 | 8 | 1161 | XP_006712461.1 | |
| XM_006712400.3 | 9 | 1239 | XP_006712463.1 | |
| XM_011510926.2 | 8 | 940 | XP_011509228.1 | |
| XM_011510928.2 | 6 | 873 | XP_011509230.1 | |
| XM_017003767.1 | 8 | 1011 | XP_016859256.1 |
Target Gene Details
Entrez Gene ID: | 90134 |
Gene Name: | potassium voltage-gated channel subfamily H member 7 |
Gene Aliases: |
ERG3, HERG3, Kv11.3 |
Location: |
Chr.2:162371407-162838747 on Build GRCh38 |
Assay Gene Location: | Within Exon 17 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNH7 | NM_033272.3 | 16 | 4159 | NP_150375.2 |
| AF032897.1 | 16 | 4030 | AAD01946.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv583520 | Chr.2:162267541 - 162611723 on Build GRCh38 | Loss |
|
| nsv1160878 | Chr.2:162355987 - 162425533 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map