Assay Details
Target Gene Details
Entrez Gene ID: | 219402 |
Gene Name: | mitochondrial translational initiation factor 3 |
Gene Aliases: |
IF3mt |
Location: |
Chr.13:27435639-27450900 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MTIF3 | NM_001166261.1 | 3 | 493 | NP_001159733.1 |
| NM_001166262.1 | 3 | 563 | NP_001159734.1 | |
| NM_001166263.1 | 4 | 592 | NP_001159735.1 | |
| NM_152912.4 | 3 | 531 | NP_690876.3 | |
| XM_006719771.3 | 4 | 579 | XP_006719834.1 | |
| XM_006719772.3 | 3 | 810 | XP_006719835.1 | |
| XM_011534957.2 | 4 | 1053 | XP_011533259.1 | |
| XM_011534958.2 | 4 | 837 | XP_011533260.1 | |
| XM_011534959.2 | 4 | 1002 | XP_011533261.1 | |
| XM_011534960.2 | 4 | 667 | XP_011533262.1 | |
| XM_011534961.2 | 4 | 1255 | XP_011533263.1 | |
| XM_011534962.2 | 4 | 832 | XP_011533264.1 | |
| XM_011534963.2 | 4 | 1420 | XP_011533265.1 | |
| XM_017020417.1 | 3 | 437 | XP_016875906.1 | |
| AF410851.1 | 2 | 398 | AAL04150.1 | |
| AK094420.1 | 1 | 1591 | ||
| BC039599.1 | 3 | 484 | ||
| BC046166.1 | 3 | 503 | AAH46166.1 | |
| BG617536.1 | 3 | 532 | ||
| BI460170.1 | 4 | 586 | ||
| BM548061.1 | 2 | 456 | ||
| BM768315.1 | ||||
| CN272769.1 | 4 | 516 | ||
| DB470026.1 |
More Information
Set Membership: |
|

Genomic Map