Assay Details
Target Gene Details
Entrez Gene ID: | 100506321 |
Gene Name: | uncharacterized LOC100506321 |
Gene Aliases: |
- |
Location: |
Chr.14:65089918-65094311 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LOC100506321 | NR_045122.1 | |||
| BI825681.1 | ||||
| DB082809.1 |
Target Gene Details
Entrez Gene ID: | 4149 |
Gene Name: | MYC associated factor X |
Gene Aliases: |
bHLHd4 |
Location: |
Chr.14:65006101-65102695 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832816 | Chr.14:65006526 - 65177927 on Build GRCh38 | Gain |
|
| esv2748779 | Chr.14:63663324 - 66595809 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map