Assay Details
Target Gene Details
Entrez Gene ID: | 92106 |
Gene Name: | oxidoreductase NAD binding domain containing 1 |
Gene Aliases: |
- |
Location: |
Chr.3:16265160-16386979 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
More Information
Set Membership: |
|

Genomic Map