Assay Details
Target Gene Details
Entrez Gene ID: | 8763 |
Gene Name: | CD164 molecule |
Gene Aliases: |
DFNA66, MGC-24, MUC-24, endolyn |
Location: |
Chr.6:109366514-109382559 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CD164 | NM_001142401.1 | 5 | 2152 | NP_001135873.1 |
| NM_001142402.1 | 5 | 2134 | NP_001135874.1 | |
| NM_001142403.1 | 7 | 1595 | NP_001135875.1 | |
| NM_001142404.1 | 6 | 1499 | NP_001135876.1 | |
| NM_006016.4 | 6 | 2191 | NP_006007.2 | |
| AF299341.1 | 6 | 2010 | AAG53906.1 | |
| AF299342.1 | 5 | 2031 | AAG53907.1 | |
| AF299343.1 | 5 | 2049 | AAG53908.1 | |
| AK301692.1 | 6 | 1229 | ||
| BC011522.2 | 6 | 2091 | AAH11522.1 | |
| D14043.1 | 7 | 1493 | BAA03130.1 | |
| EB388947.1 | 1 | 686 |
More Information
Set Membership: |
|

Genomic Map