Assay Details
Target Gene Details
Entrez Gene ID: | 10660 |
Gene Name: | ladybird homeobox 1 |
Gene Aliases: |
HPX-6, HPX6, LBX1H, homeobox |
Location: |
Chr.10:101226976-101228960 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LBX1 | NM_006562.4 | 2 | 651 | NP_006553.2 |
| AJ004949.1 | CAA06237.1 | |||
| BC069156.1 | 2 | 652 | ||
| BC136321.1 | 2 | 623 | ||
| X74863.1 | CAA52856.1 | |||
| X90828.1 | 2 | 644 | CAA62342.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv831966 | Chr.10:101124857 - 101287432 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map