Assay Details
Target Gene Details
Entrez Gene ID: | 158 |
Gene Name: | adenylosuccinate lyase |
Gene Aliases: |
AMPS, ASASE, ASL |
Location: |
Chr.22:40346500-40387408 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADSL | NM_000026.3 | NP_000017.1 | ||
| NM_001123378.2 | NP_001116850.1 | |||
| NM_001317923.1 | NP_001304852.1 | |||
| NR_134256.1 | ||||
| XM_011529977.2 | XP_011528279.1 | |||
| XM_011529979.1 | XP_011528281.1 | |||
| XM_011529980.2 | XP_011528282.1 | |||
| XM_017028636.1 | XP_016884125.1 | |||
| XM_017028637.1 | XP_016884126.1 | |||
| XM_017028638.1 | XP_016884127.1 | |||
| XM_017028639.1 | XP_016884128.1 | |||
| AF067853.1 | AAC21560.1 | |||
| AF067854.1 | AAC21561.1 | |||
| AK225042.1 | ||||
| AK291091.1 | ||||
| AK293722.1 | ||||
| AK298187.1 | ||||
| AK300705.1 | ||||
| BC000253.1 | AAH00253.1 | |||
| BP298407.1 | ||||
| CR456368.1 | CAG30254.1 | |||
| KJ534763.1 | ||||
| KJ535007.1 | ||||
| KJ535020.1 | ||||
| KJ535041.1 | ||||
| KJ535056.1 | ||||
| S60710.1 | AAC60603.1 | |||
| X65867.1 | CAA46696.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834202 | Chr.22:40179762 - 40385241 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map