Assay Details
Target Gene Details
Entrez Gene ID: | 134526 |
Gene Name: | acyl-CoA thioesterase 12 |
Gene Aliases: |
CACH-1, Cach, STARD15, THEAL |
Location: |
Chr.5:81309045-81394174 on Build GRCh38 |
Assay Gene Location: | Within Intron 16 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ACOT12 | XM_017009045.1 | XP_016864534.1 | ||
| XM_017009046.1 | XP_016864535.1 | |||
| XM_017009047.1 | XP_016864536.1 | |||
| XM_017009048.1 | XP_016864537.1 |
Target Gene Details
Entrez Gene ID: | 84240 |
Gene Name: | zinc finger CCHC-type containing 9 |
Gene Aliases: |
PPP1R41 |
Location: |
Chr.5:81301583-81313147 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZCCHC9 | NM_001131035.1 | NP_001124507.1 | ||
| NM_001131036.1 | NP_001124508.1 | |||
| NM_032280.2 | NP_115656.1 | |||
| XM_005248616.3 | XP_005248673.1 | |||
| AK314160.1 | ||||
| AL512712.1 | CAC21654.1 | |||
| BC032736.1 | AAH32736.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3890664 | Chr.5:81194463 - 81323227 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map