Assay Details
Target Gene Details
Entrez Gene ID: | 51241 |
Gene Name: | COX16, cytochrome c oxidase assembly homolog |
Gene Aliases: |
C14orf112, HSPC203 |
Location: |
Chr.14:70325081-70359731 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COX16 | NM_001204090.1 | 1 | 78 | NP_001191019.1 |
| NM_016468.6 | 1 | 78 | NP_057552.1 | |
| AF151037.1 | AAF36123.1 | |||
| AF226729.1 | AAG09730.1 | |||
| AK290743.1 | ||||
| BC001702.2 | AAH01702.1 | |||
| BM799209.1 | ||||
| CB156527.1 | ||||
| FJ460510.1 | 1 | 78 | ||
| FJ460511.1 | 1 | 78 |
Target Gene Details
Entrez Gene ID: | 100529257 |
Gene Name: | SYNJ2BP-COX16 readthrough |
Gene Aliases: |
- |
Location: |
Chr.14:70325081-70417090 on Build GRCh38 |
Assay Gene Location: | Within Intron 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SYNJ2BP-COX16 | NM_001202547.2 | NP_001189476.1 | ||
| NM_001202548.2 | NP_001189477.1 | |||
| NM_001202549.2 | NP_001189478.1 | |||
| FY210787.1 | ||||
| FY211163.1 | ||||
| FY213292.1 | ||||
| FY213836.1 |
More Information
Set Membership: |
|

Genomic Map