Assay Details
Target Gene Details
Entrez Gene ID: | 5734 |
Gene Name: | prostaglandin E receptor 4 |
Gene Aliases: |
EP4, EP4R |
Location: |
Chr.5:40679930-40740932 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGER4 | NM_000958.2 | 3 | 2852 | NP_000949.1 |
| XM_017009659.1 | XP_016865148.1 | |||
| BQ029720.1 | ||||
| CD000197.1 | 1 | 369 |
More Information
Set Membership: |
|

Genomic Map