Assay Details
Target Gene Details
Entrez Gene ID: | 57369 |
Gene Name: | gap junction protein delta 2 |
Gene Aliases: |
CX36, GJA9 |
Location: |
Chr.15:34751746-34754955 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GJD2 | NM_020660.2 | NP_065711.1 | ||
| XM_017022438.1 | XP_016877927.1 | |||
| BC069339.1 | AAH69339.1 | |||
| BC112110.1 | AAI12111.1 | |||
| BC112114.1 | AAI12115.1 | |||
| BC143788.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv569133 | Chr.15:34589103 - 34850365 on Build GRCh38 | Gain |
|
| esv3569431 | Chr.15:33564570 - 35080307 on Build GRCh38 | Gain |
|
| nsv522411 | Chr.15:34661785 - 35170780 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map