Assay Details
Target Gene Details
Entrez Gene ID: | 164045 |
Gene Name: | HFM1, ATP dependent DNA helicase homolog |
Gene Aliases: |
MER3, POF9, SEC63D1, Si-11, Si-11-6, helicase |
Location: |
Chr.1:91260766-91408008 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 30 - Exon 30 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HFM1 | NM_001017975.4 | NP_001017975.4 | ||
| XM_011540849.1 | XP_011539151.1 | |||
| XM_011540850.2 | XP_011539152.1 | |||
| XM_011540851.1 | XP_011539153.1 | |||
| XM_011540852.2 | XP_011539154.1 | |||
| XM_011540855.1 | XP_011539157.1 | |||
| XM_011540857.1 | XP_011539159.1 | |||
| XM_011540859.2 | XP_011539161.1 | |||
| XM_017000490.1 | XP_016855979.1 | |||
| XM_017000491.1 | XP_016855980.1 | |||
| XM_017000492.1 | XP_016855981.1 | |||
| XM_017000493.1 | XP_016855982.1 | |||
| XM_017000494.1 | XP_016855983.1 | |||
| AB044787.1 | ||||
| AB204867.1 | ||||
| AK094079.1 | BAC04281.1 | |||
| AK294754.1 | ||||
| AK302582.1 | ||||
| BC144172.1 | ||||
| BX649037.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv546806 | Chr.1:91314851 - 91409948 on Build GRCh38 | Loss |
|
| nsv1006022 | Chr.1:91303083 - 91365639 on Build GRCh38 | Loss |
|
| esv2714684 | Chr.1:91095800 - 93049130 on Build GRCh38 | Deletion |
|
More Information
Additional Information:
For this assay, SNP(s) [rs74843031] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map